InocuLoop Inoculating Loops, Caplugs
InocuLoop Inoculating Loops, Caplugs
Loop and spreader, 10 µl, ABS, Light yellow
Catalog # 76492-640
Supplier:   Caplugs
Supplier #:  333-5010-Y10
InocuLoop Inoculating Loops, Caplugs
Catalog # 76492-640
Supplier:  Caplugs
InocuLoop Inoculating Loops, Caplugs
InocuLoop Inoculating Loops, Caplugs
Loop and spreader, 10 µl, ABS, Light yellow
Price
$141.42
Log in pour la disponibilité
Carton of 500
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Inoculoop disposable inoculating loops have patented four-sided spreaders, enabling the inoculation of a petri dish with one, not multiple, disposable loops. Make bacterial inoculation and quantitative procedures faster and more efficient.
  • Made of flexible plastic for easy spreading
  • Can also be used as a picker for isolating colonies
  • Pre-sterilized Inoculoops are gamma-irritated
Suitable in anaerobic chambers, in the field and even in satellite metal loops.
Spécifications
  • Capacité:
    10 µl
  • Color:
    Light yellow
  • Désignation:
    Loop and spreader
  • Matière:
    ABS