Dissection Probes
Angled probe and seeker, Stainless steel, 6", Blunt
Catalog # 470334-892
Supplier: AFTAB AHMED SE
Fournisseur #: 14-0950
Jump to:
Product Details & Documents
Easily tease up fine tissues.
- Blunt tips prevent accidental tissue damage
- Stainless steel
Probes provide an easy way to lift embedded tissue, or track the path of nerves and vessels.
The spear point is sharp to allow controlled removal of connective tissue, while the blunt probes allow for easily lifting tissue and elevating.
Blunt probes are also very good for breaking through loose connective tissue without damaging other tissues.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Angled probe and seeker
- Longueur:6"
- Matière:Stainless steel
- Tip Style:Blunt