Dissection Knives | Avantor
Dissection Knives
Cartilage knife, Stainless steel, 6.5"
Catalog # 470334-890
Supplier:   AFTAB AHMED SE
Fournisseur #:  14-0930
Dissection Knives
Catalog # 470334-890
Supplier:  AFTAB AHMED SE
Dissection Knives
Dissection Knives
Cartilage knife, Stainless steel, 6.5"
Price
$10.50
Log in pour la disponibilité
Each Box of 12
Product Details & Documents
Great for rough dissection.
  • Stainless steel construction
  • Fixed blades for safety
More durable than scalpels, dissection knives are great for rough dissection, rapid fleshing, or dealing with tough tissues. The basic fleshing knife has a shorter blade and is easily controlled.
The Amputation knife has an extended blade for long smooth cuts through soft tissues. Use the cartilage knife, which has a scalpel form, to easily reach into tight areas to cut tough tissue.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
  • Désignation:
    Cartilage knife
  • Longueur:
    6.5"
  • Matière:
    Stainless steel