Dissection Scissors | Avantor
Dissection Scissors
Scissors straight blade, 5.5", Stainless steel, Sharp/sharp
Catalog # 470334-878
Supplier:   AFTAB AHMED SE
Fournisseur #:  10-917
Dissection Scissors
Catalog # 470334-878
Supplier:  AFTAB AHMED SE
Dissection Scissors
Dissection Scissors
Scissors straight blade, 5.5", Stainless steel, Sharp/sharp
Price
$12.55
Log in pour la disponibilité
Each Box of 12
Product Details & Documents
Basic dissection for students.
  • Stainless steel
Basic dissection scissors are strong, durable, and have shorter handles for good control. A good option for inexperienced students, these have a slightly blunted pointed blade set to also minimize the chance for accidental pokes.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
  • Désignation:
    Scissors straight blade
  • Longueur:
    5.5"
  • Matière:
    Stainless steel
  • Blade Type:
    Sharp/sharp