Iris Scissors
Micro iris, 4.75", Stainless steel, Sharp/sharp
Catalog # 470334-872
Supplier: AFTAB AHMED SE
Supplier #: 14-0976
Jump to:
Product Details & Documents
Extremely fine scissors.
- Stainless steel
Double sharp points, a very fine blade thickness, and shorter handles make Iris scissors the easiest to control in very tight locations. The blades will very cleanly cut even very fine tissue.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Micro iris
- Longueur:4.75"
- Matière:Stainless steel
- Blade Type:Sharp/sharp