Dissection Scissors
Mayo scissors, 5.5", Stainless steel, Sharp/sharp
Catalog # 470334-870
Supplier: AFTAB AHMED SE
: 10-916
Jump to:
Product Details & Documents
Basic dissection for students.
- Stainless steel
Basic dissection scissors are strong, durable, and have shorter handles for good control. A good option for inexperienced students, these have a slightly blunted pointed blade set to also minimize the chance for accidental pokes.
: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Mayo scissors
- Longueur:5.5"
- Matière:Stainless steel
- Blade Type:Sharp/sharp