Iris Scissors | Avantor
Iris Scissors
Iris scissors, 4.5", Tungsten carbide, Sharp/sharp
Catalog # 470334-868
Supplier:   AFTAB AHMED SE
Fournisseur #:  10-901
Iris Scissors
Catalog # 470334-868
Supplier:  AFTAB AHMED SE
Iris Scissors
Iris Scissors
Iris scissors, 4.5", Tungsten carbide, Sharp/sharp
Price
38,35 $
Log in pour la disponibilité
Each Box of 12
Product Details & Documents
Extremely fine scissors.
  • Stainless steel
Double sharp points, a very fine blade thickness, and shorter handles make Iris scissors the easiest to control in very tight locations. The blades will very cleanly cut even very fine tissue.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
  • Désignation:
    Iris scissors
  • Longueur:
    4.5"
  • Matière:
    Tungsten carbide
  • Blade Type:
    Sharp/sharp