Dissection Knives
Liston amputation knife, Stainless steel, 8"
Catalog # 470334-852
Supplier: AFTAB AHMED SE
Fournisseur #: 14-0969
Jump to:
Product Details & Documents
Great for rough dissection.
- Stainless steel construction
- Fixed blades for safety
More durable than scalpels, dissection knives are great for rough dissection, rapid fleshing, or dealing with tough tissues. The basic fleshing knife has a shorter blade and is easily controlled.
The Amputation knife has an extended blade for long smooth cuts through soft tissues. Use the cartilage knife, which has a scalpel form, to easily reach into tight areas to cut tough tissue.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Liston amputation knife
- Longueur:8"
- Matière:Stainless steel