Dissection Knives
Fleshing knife, Stainless steel, 7"
  470334-844
 :  14-1190
Dissection Knives
  470334-844
Dissection Knives
Dissection Knives
Fleshing knife, Stainless steel, 7"
$19.20
Each Box of 12
Great for rough dissection.
  • Stainless steel construction
  • Fixed blades for safety
More durable than scalpels, dissection knives are great for rough dissection, rapid fleshing, or dealing with tough tissues. The basic fleshing knife has a shorter blade and is easily controlled.
The Amputation knife has an extended blade for long smooth cuts through soft tissues. Use the cartilage knife, which has a scalpel form, to easily reach into tight areas to cut tough tissue.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
  • Désignation:
    Fleshing knife
  • Longueur:
    7"
  • Matière:
    Stainless steel