Dissection Knives
Fleshing knife, Stainless steel, 7"
470334-844
: 14-1190
Great for rough dissection.
- Stainless steel construction
- Fixed blades for safety
More durable than scalpels, dissection knives are great for rough dissection, rapid fleshing, or dealing with tough tissues. The basic fleshing knife has a shorter blade and is easily controlled.
The Amputation knife has an extended blade for long smooth cuts through soft tissues. Use the cartilage knife, which has a scalpel form, to easily reach into tight areas to cut tough tissue.
: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
- Désignation:Fleshing knife
- Longueur:7"
- Matière:Stainless steel