Toothed Forceps
Toothed Forceps
Tissue forceps, Stainless steel, 4.5", Straight
Catalog # 470334-842
Supplier:   AFTAB AHMED SE
Supplier #:  14-1015
Toothed Forceps
Catalog # 470334-842
Supplier:  AFTAB AHMED SE
Toothed Forceps
Toothed Forceps
Tissue forceps, Stainless steel, 4.5", Straight
Price
$1.40
Log in for availability
Each Box of 50
Product Details & Documents
Toothed grip for a secure hold.
  • Stainless steel
These stainless steel dissection forceps are great for general usage. The variety of tip sizes and shapes accommodates a wide range of application specific needs.
These all have a toothed jaw to provide a very secure grip on tough or slippery tissue.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Désignation:
    Tissue forceps
  • Longueur:
    4.5"
  • Matière:
    Stainless steel
  • Tip Style:
    Straight