Toothed Forceps
Gilles forceps, Stainless steel, 4.5", Straight
Catalog # 470334-838
Supplier: AFTAB AHMED SE
: 14-1017
Jump to:
Product Details & Documents
Toothed grip for a secure hold.
- Stainless steel
These stainless steel dissection forceps are great for general usage. The variety of tip sizes and shapes accommodates a wide range of application specific needs.
These all have a toothed jaw to provide a very secure grip on tough or slippery tissue.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Gilles forceps
- Longueur:4.5"
- Matière:Stainless steel
- Tip Style:Straight