Serrated Forceps
Serrated Forceps
Stainless steel, 4.5", Bent
Catalog # 470334-836
Supplier:   AFTAB AHMED SE
Supplier #:  14-1009
Serrated Forceps
Catalog # 470334-836
Supplier:  AFTAB AHMED SE
Serrated Forceps
Serrated Forceps
Stainless steel, 4.5", Bent
Price
$2.30
Log in for availability
Each Box of 50
Product Details & Documents
Serrated grips for less tissue damage.
  • Stainless steel
These stainless steel dissection forceps are great for general usage. The variety of tip sizes and shapes accommodates a wide range of application specific needs.
These all feature a serrated jaw to provide a secure grip without causing significant tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Longueur:
    4.5"
  • Matière:
    Stainless steel
  • Tip Style:
    Bent