Dissection Needles
Needle for needle holder, -, -, -
Catalog # 470334-822
Supplier: AFTAB AHMED SE
Fournisseur #: 14-0633
Jump to:
Product Details & Documents
Sharp point needles.
- Fine point teasing needles
- Straight or angled tips
Very sharp points on these needles make tissue separation fast and easy. Used mostly to break apart muscle tissues.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Needle for needle holder
- Longueur:-
- Matière:-
- Tip Style:-