Hemostatic Forceps
Hemostatic forceps, Stainless steel, 5.5", Curved
Catalog # 470334-816
Fournisseur #: 20-105
Jump to:
Discontinued
Product Details & Documents
Lock for a secure hold.
- Stainless steel
- Serrated jaws
Hemostatic forceps are a locking forceps, with thumb loops that operate much like scissors. Often referred to as clamps, these have a serrated jaw to provide a secure hold while minimizing tissue damage.
: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Désignation:Hemostatic forceps
- Longueur:5.5"
- Matière:Stainless steel
- Tip Style:Curved