Hemostatic Forceps | Avantor
Hemostatic Forceps
Hemostatic forceps, Stainless steel, 5.5", Curved
Catalog # 470334-816
Fournisseur #:  20-105
Hemostatic Forceps
Catalog # 470334-816
Supplier:  
Hemostatic Forceps
Hemostatic Forceps
Hemostatic forceps, Stainless steel, 5.5", Curved
Discontinued
Product Details & Documents
Lock for a secure hold.
  • Stainless steel
  • Serrated jaws
Hemostatic forceps are a locking forceps, with thumb loops that operate much like scissors. Often referred to as clamps, these have a serrated jaw to provide a secure hold while minimizing tissue damage.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
  • Désignation:
    Hemostatic forceps
  • Longueur:
    5.5"
  • Matière:
    Stainless steel
  • Tip Style:
    Curved