Special Use Forceps | Avantor
Special Use Forceps
Self closing, Stainless steel, 4.5", Straight
Catalog # 470334-814
Supplier:   AFTAB AHMED SE
Fournisseur #:  15-122
Special Use Forceps
Catalog # 470334-814
Supplier:  AFTAB AHMED SE
Special Use Forceps
Special Use Forceps
Self closing, Stainless steel, 4.5", Straight
Price
$6.90
Log in pour la disponibilité
Each Box of 12
Product Details & Documents
Specialty forceps for student use.
  • Stainless steel
  • A variety of options available
One set of forceps isn't like every other. This selection provides options for many users. Tweezers are a basic sharp point forceps with a very short blade length that makes the ends very easy to control and provides a stronger grip.
Cover slipping forceps are designed with a broader smooth tip that is bent at an angle to improve applying a coverslip to a microscope slide.
Microdissection forceps are similar to a watchmakers forceps with a lighter weight blade, requiring less force to close. Easy grip forceps have an enlarged grip loop part-way down each blade to provide additional stability and ease of closure for a narrow blade.
Finally the self closing forceps act in a reverse motion to most. Squeezing opens the jaws, and they close when released. These are easier to use if long-term holding is required.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
  • Désignation:
    Self closing
  • Longueur:
    4.5"
  • Matière:
    Stainless steel
  • Tip Style:
    Straight
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.