Watchmaker Forceps
Stainless steel, 4", Straight
Catalog # 470334-802
Supplier: AFTAB AHMED SE
Fournisseur #: 14-1400
Jump to:
Product Details & Documents
Ultra-fine point forceps.
- Stainless steel
Extremely fine tips and heavier handles allow for the manipulation of very fine tissue samples with a secure hold.
These forceps have a smooth finish inside the jaws, to ensure a minimum of tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
- Longueur:4"
- Matière:Stainless steel
- Tip Style:Straight