Watchmaker Forceps | Avantor
Watchmaker Forceps
Stainless steel, 4", Straight
Catalog # 470334-802
Supplier:   AFTAB AHMED SE
Fournisseur #:  14-1400
Watchmaker Forceps
Catalog # 470334-802
Supplier:  AFTAB AHMED SE
Watchmaker Forceps
Watchmaker Forceps
Stainless steel, 4", Straight
Price
$9.65
Log in pour la disponibilité
Each Box of 12
Product Details & Documents
Ultra-fine point forceps.
  • Stainless steel
Extremely fine tips and heavier handles allow for the manipulation of very fine tissue samples with a secure hold.
These forceps have a smooth finish inside the jaws, to ensure a minimum of tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
  • Longueur:
    4"
  • Matière:
    Stainless steel
  • Tip Style:
    Straight