Hemostatic Forceps
Halstead mosquito clamp, Stainless steel, 5", Straight
Catalog # 470334-792
Supplier: AFTAB AHMED SE
Supplier #: 14-0551
Jump to:
Product Details & Documents
Lock for a secure hold.
- Stainless steel
- Serrated jaws
Hemostatic forceps are a locking forceps, with thumb loops that operate much like scissors. Often referred to as clamps, these have a serrated jaw to provide a secure hold while minimizing tissue damage.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Désignation:Halstead mosquito clamp
- Longueur:5"
- Matière:Stainless steel
- Tip Style:Straight