Serrated Forceps
Stainless steel, 4.5", Straight
Catalog # 470334-786
Supplier: AFTAB AHMED SE
Supplier #: 14-1004
Jump to:
Product Details & Documents
Serrated grips for less tissue damage.
- Stainless steel
These stainless steel dissection forceps are great for general usage. The variety of tip sizes and shapes accommodates a wide range of application specific needs.
These all feature a serrated jaw to provide a secure grip without causing significant tissue damage.
: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
- Longueur:4.5"
- Matière:Stainless steel
- Tip Style:Straight