Toothed Forceps
Toothed Forceps
Adison brown, Stainless steel, 4.5", Straight
Catalog # 470334-768
Supplier:   AFTAB AHMED SE
Supplier #:  15-103
Toothed Forceps
Catalog # 470334-768
Supplier:  AFTAB AHMED SE
Toothed Forceps
Toothed Forceps
Adison brown, Stainless steel, 4.5", Straight
Price
$11.10
Log in pour la disponibilité
Each Box of 12
Product Details & Documents
Toothed grip for a secure hold.
  • Stainless steel
These stainless steel dissection forceps are great for general usage. The variety of tip sizes and shapes accommodates a wide range of application specific needs.
These all have a toothed jaw to provide a very secure grip on tough or slippery tissue.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
  • Désignation:
    Adison brown
  • Longueur:
    4.5"
  • Matière:
    Stainless steel
  • Tip Style:
    Straight