Bone Shears | Avantor
Bone Shears
Bone cutter, fine point, 5.5", Stainless steel
Catalog # 470334-764
Supplier:   AFTAB AHMED SE
Fournisseur #:  14-0995
Bone Shears
Catalog # 470334-764
Supplier:  AFTAB AHMED SE
Bone Shears
Bone Shears
Bone cutter, fine point, 5.5", Stainless steel
Price
$51.75
Log in pour la disponibilité
Each Box of 6
Product Details & Documents
Easily snip through bones.
  • Stainless steel
  • Flush cutting face
  • Short jaws for additional force
Used to easily cut through tough tissue or bone during dissection. These bone cutting shears combine longer grips and shorter heads to make cuts easier. The cutter has a beveled edge on one side for easy flush cutting.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Spécifications
  • Désignation:
    Bone cutter, fine point
  • Longueur:
    5.5"
  • Matière:
    Stainless steel