CTL Data Loggers, Cryopak Verification Technologies
Data logger, CTL single trip in sleeve with beeper data
Catalog # 76490-416
Supplier: Cryopak Verification Technologies
Fournisseur #: CTL-ST-4-B
CTL Data Loggers, Cryopak Verification Technologies
Catalog # 76490-416
Supplier: Cryopak Verification Technologies
Jump to:
Product Details & Documents
This contactless data logger works with a corresponding mobile app to provide maximum flexiblity.
- High accuracy
- Large memory capacity
- Long battery life
Encased in a waterproof pouch and utilizing seven LED indicators, the CTL data logger is pre-programmed to customer specifications.
Spécifications
- Désignation:Data logger, CTL single trip in sleeve with beeper data