Objectives for REVOLVE Microscopes, Echo
Apochromat oil objective, 100×, 0.13 mm, 1.45 NA
Catalog # 76490-280
Supplier #: JMAG9001
Jump to:
Discontinued
Product Details & Documents
Specifications
- Grossissement:100×
- Désignation:Apochromat oil objective
- Aperture:1.45 NA
- Working Distance:0.13 mm