Objectives for REVOLVE Microscopes, Echo
Apochromat oil phase objective, 60×, 0.15 mm, 1.42 NA
Catalog # 76490-278
Fournisseur #: JMAG9602
Jump to:
Discontinued
Product Details & Documents
Spécifications
- Grossissement:60×
- Désignation:Apochromat oil phase objective
- Aperture:1.42 NA
- Working Distance:0.15 mm