Objectives for REVOLVE Microscopes, Echo
Apochromat oil objective, 60×, 0.15 mm, 1.42 NA
Catalog # 76490-276
Fournisseur #: JMAG9601
Jump to:
Discontinued
Product Details & Documents
Spécifications
- Grossissement:60×
- Désignation:Apochromat oil objective
- Aperture:1.42 NA
- Working Distance:0.15 mm