Objectives for REVOLVE Microscopes, Echo
Apochromat objective, 20×, 0.6 mm, 0.80 NA
Catalog # 76490-270
Fournisseur #: JMAG9201
Jump to:
Discontinued
Product Details & Documents
Spécifications
- Grossissement:20×
- Désignation:Apochromat objective
- Aperture:0.80 NA
- Working Distance:0.6 mm