Objectives for REVOLVE Microscopes, Echo
Apochromat objective, 10×, 3.1 mm, 0.40 NA
Catalog # 76490-268
Fournisseur #: JMAG9101
Jump to:
Discontinued
Product Details & Documents
Spécifications
- Grossissement:10×
- Désignation:Apochromat objective
- Aperture:0.40 NA
- Working Distance:3.1 mm