Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Extracta™ DNA Prep for PCR, Quantabio | Avantor

Extracta™ DNA Prep for PCR, Quantabio

Extracta™ DNA prep for PCR, 2.5 ml
Catalog # CA76047-142
Supplier:   Quantabio
Fournisseur #:  95091-002
Extracta™ DNA Prep for PCR, Quantabio
Catalog # CA76047-142
Supplier:  Quantabio
CAS Number:  
Extracta™ DNA Prep for PCR, Quantabio
Extracta™ DNA Prep for PCR, Quantabio

Extracta™ DNA prep for PCR, 2.5 ml

CAS Number:  
Price
$32.01
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Extracta DNA Prep for PCR is a two-component reagent kit for rapid extraction of PCR-ready genomic DNA from a variety of tissues. Samples are processed in less than 30 minutes with minimal hands-on time and technical skill.

  • Simple, fast protocol
  • Compatible with laboratory automation
  • DNA is suitable for qPCR, High Resolution Melt Analysis, endpoint PCR and other applications
  • Works with wide range of tissue including hair, buccal swabs and mouse tails
  • Optional 10 minute protocol for high-throughput applications

Quanta's Extracta sample preparation system is a system that can be scaled to process various amounts of solid tissue in a simple and fast single-tube procedure. DNA may be purified from many tissue types and is suitable for use in sensitive downstream applications such as PCR, qPCR and High Resolution Melt Analysis.

The Extracta™ DNA prep for PCR consists of a two component reagent kit for rapid and efficient extraction of PCR-ready genomic DNA from mammalian tissues. The kit has been demonstrated to work with mouse tail biopsies, ear punches and a variety of human tissue samples including hair, buccal cells and saliva. The genomic DNA is suitable for sensitive downstream PCR applications including end-point PCR, High Resolution Melt Analysis (HRM) and quantitative real time PCR (qPCR) without requiring any additional clean-up. In addition, the extracted DNA may be used in multiplexed PCR applications such as transgene or knock-out analyses. Tissue extractions can be performed in tubes, plates or deep well blocks to allow for adaptation to workflow and automation on liquid handling workstations.

Ordering information: DNA Prep kit includes extraction reagent and stabilization buffer.

Documents

Spécifications

Spécifications

  • Désignation:
    Extracta™ DNA prep for PCR
  • Taille:
    2.5 ml
  • Cat. No.:
    CA76047-142
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Extraction for direct PCR
  • Description:
    Extracta™ DNA prep for PCR
  • Sample type:
    Cultured cells|Human/animal cells and tissues

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.