TraceableLive® Subscription Coupon Codes
TraceableLive® Subscription Coupon Codes
TraceableLive® 1 year standard subscription coupon code
Catalog # 76488-212
Supplier:   Antyila Scientific
Supplier #:  63003-36
TraceableLive® Subscription Coupon Codes
Catalog # 76488-212
Supplier:  Antyila Scientific
TraceableLive® Subscription Coupon Codes
TraceableLive® Subscription Coupon Codes
TraceableLive® 1 year standard subscription coupon code
Price
$210.61
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Simplified cloud monitoring.
  • TraceableLive®-capable products securely monitor critical environments and notify smartphones, tablets, or PCs if anything goes wrong
  • Notifications for peace of mind
  • All team members can receive email, text, and push notifications on the environments monitored
  • Remotely view data logging history, effortlessly generate reports, and control alarm parameters
  • Traceable products come calibrated to NIST standards to meet quality and regulatory requirements
  • Digital data reports sent with ease whenever needed
  • Customize reports with location, device, date range, and report format
  • No limit to the number of devices on a TraceableLive® account for the Standard and Premium plans
Purchase a subscription code for one or two years of TraceableLive®. Subscriptions are available in standard or premium formats and are required per device. Scan the QR code to begin setup or go to traceablelive.com/activate and enter the unique 11 digit number on the card.
A standard subscription includes the following features: E-mail and Push Alarms, Unlimited Data Storage, Unlimited Team Members, Calibration Due notification, SMS (Text) - Not included with the subscription code, add on in the TraceableLive® app.
A premium subscription includes the following features: E-mail and Push Alarms, Free SMS (Text) Notifications, Unlimited Data Storage, Unlimited Team Members, Calibration Due notification, 21 CFR Part 11 Compliance.
Spécifications
  • Désignation:
    TraceableLive® 1 year standard subscription coupon code