Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Disposable Transfer Pipettes

Disposable transfer pipette, 3 ml, 140 mm
  470334-416
 :  25013
Disposable Transfer Pipettes
  470334-416
Disposable Transfer Pipettes
Disposable Transfer Pipettes

Disposable transfer pipette, 3 ml, 140 mm

 
$37.45
   
Pack of 500 (3ml)
 

Used in many applications such as urinalysis, wet chemistry, microbiology, hematology, and serology.

  • Small micro-tip
  • Made from LDPE

Ensure a clean, contamination-free experience every time. Whether working in a lab, a medical facility, or an industrial setting, this is an essential tool for STEM professionals.

 
  • Volume:
    3 ml
  • Désignation:
    Disposable transfer pipette
  • Longueur:
    140 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.