Disposable Transfer Pipettes
Disposable transfer pipette, 5 ml, 152.4 mm
Catalog # 470334-406
Supplier: GSC International
Supplier #: 25008
Jump to:
Product Details & Documents
Used in many applications such as urinalysis, wet chemistry, microbiology, hematology, and serology.
- Small micro-tip
- Made from LDPE
Ensure a clean, contamination-free experience every time. Whether working in a lab, a medical facility, or an industrial setting, this is an essential tool for STEM professionals.
Specifications
- Volume:5 ml
- Désignation:Disposable transfer pipette
- Longueur:152.4 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



