SmartCheck DNA Ladders
100 bp, 5× 500 µl, 500
470334-114
: Ward's
Supplier #: PR4010-500
SmartCheck ladders are available in three size ranges (50 bp, 100 bp and 1 kb) to cover most common applications.
- Ready-to-use formulation includes loading buffer and tracking dye
- 500 µl suitable for 100 lanes (5 µl per lane)
- Higher intensity reference bands
- Ultra pure production allows economical ambient shipping
- Taille:100 bp
- Volume:5× 500 µl
- Lanes:500