SmartCheck DNA Ladders
SmartCheck DNA Ladders
100 bp, 5× 500 µl, 500
  470334-114
 :   Ward's
Supplier #:  PR4010-500
SmartCheck DNA Ladders
  470334-114
 :  Ward's
 :  
SmartCheck DNA Ladders
SmartCheck DNA Ladders
100 bp, 5× 500 µl, 500
 :  
$326.75
Each
SmartCheck ladders are available in three size ranges (50 bp, 100 bp and 1 kb) to cover most common applications.
  • Ready-to-use formulation includes loading buffer and tracking dye
  • 500 µl suitable for 100 lanes (5 µl per lane)
  • Higher intensity reference bands
  • Ultra pure production allows economical ambient shipping
  • Taille:
    100 bp
  • Volume:
    5× 500 µl
  • Lanes:
    500