Unisette™ Tissue Processing and Biopsy Cassettes, Simport
Unisette™ Tissue Processing and Biopsy Cassettes, Simport
Pink, Case of 1000
Catalog # 76479-316
Supplier:   Simport Scientific
Supplier #:  M526-3T
Unisette™ Tissue Processing and Biopsy Cassettes, Simport
Catalog # 76479-316
Supplier:  Simport Scientific
Unisette™ Tissue Processing and Biopsy Cassettes, Simport
Unisette™ Tissue Processing and Biopsy Cassettes, Simport
Pink, Case of 1000
Price
$276.85
Log in pour la disponibilité
Case of 1,000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our extremely popular Unisette™ histology cassette is now offered with a 45° anterior writing surface. This configuration is considered ideal for optimal workflow in some automated cassette printers and efficient barcode reading.
  • Made of acetal
  • 45° anterior writing area
  • One-piece integral lid that can be opened and closed up to three times without breakage
  • Secure locking system without danger of specimen loss
  • The biopsy cassette has 1 mm square openings to maximize fluid exchange
  • Offered taped for use with Leica™ IPC printers
Molded from acetal, these cassettes keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The snap latch and hinge-lock design of the UNISETTE™ prevent early separation of base and lid and allows for one-hand operation.
Spécifications
  • Color:
    Pink
  • Units:
    Case of 1000