Plastic Beakers
100 ml, 10 ml
Catalog # 470332-660
Supplier: United Scientific Supplies
Fournisseur #: BS100-PK/12
Jump to:
Product Details & Documents
Plastic beakers for classroom activites.
- Raised graduations
- Easy pour spout
These plastic beakers are perfect additions to your classroom. The graduations are easy to read.
Spécifications
- Intervalle de graduation:10 ml
- Capacité:100 ml