Polyethylene Dropping Bottles
473 ml (16 oz.), LDPE
470332-528
: Dynalon
: 605065-0160
Economy LDPE dropper bottles.
- Translucent LDPE dropper bottles
- Sturdy white dropping cap
- Flip up spout
Dropper bottles with hinged dispenser caps for dispensing indicator solutions, dyes, and more.
- Capacité:473 ml (16 oz.)
- Matière:LDPE