Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mini Plant Press | Avantor

Mini Plant Press

Blotter dryer sheet, 9", 6"
Catalog # 470332-442
Supplier:   Ward's Science
Fournisseur #:  506.4021.100
Mini Plant Press
Catalog # 470332-442
Supplier:  Ward's Science
Mini Plant Press
Mini Plant Press

Blotter dryer sheet, 9", 6"

Price
$67.00
Log in pour la disponibilité
Pack of 100
Product Details & Documents

For hobbyist or field collection.

  • Sized for easy portability
  • Elastic bands provide even compression

The solution to the field ecologist's or naturalist's problem of bringing back specimens for identification from the field. These very portable yet extremely functional mini-presses should be in every field biologists kit. As a teaching tool and for student use the mini-press is ideal.

Delivery information: Plant press has two masonite boards, 12 cards and driers and two elastic bands.

Spécifications
  • Désignation:
    Blotter dryer sheet
  • Longueur:
    9"
  • Width:
    6"
  • Cat. No.:
    470332-442