Fischertechnik TXT 4.0 Base Set | Avantor
Fischertechnik TXT 4.0 Base Set
Fischertechnik TXT 4.0 omniwheels
Catalog # 470332-598
Supplier:   STUDICA INC.
Fournisseur #:  559898
Fischertechnik TXT 4.0 Base Set
Catalog # 470332-598
Supplier:  STUDICA INC.
Fischertechnik TXT 4.0 Base Set
Fischertechnik TXT 4.0 Base Set
Fischertechnik TXT 4.0 omniwheels
Price
$485.50
Log in pour la disponibilité
Each
Product Details & Documents
Measurement, control, regulation, and programming for middle/high school and beyond.
  • Acts as the core set for all fischertechnik robotic add-on products
  • Includes downloadable teaching and activity material (free download)
Main topics include robotics and programming (graphical and text-based), actuators, analog and digital sensors, measurement, control, data transmission, coding-decoding, and image processing. Set included a robotics TXT 4.0 controller, robo pro coding software, accu set, 2 encoder motors, USB camera, ultrasonic sensor, track sensor, 2 push button switches, 2 LEDs, and a photo-transistor.
Spécifications
  • Désignation:
    Fischertechnik TXT 4.0 omniwheels
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.