Fischertechnik TXT 4.0 Base Set | Avantor
Fischertechnik TXT 4.0 Base Set
Fischertechnik TXT 4.0 base set
Catalog # 470332-592
Fournisseur #:  559888
Fischertechnik TXT 4.0 Base Set
Catalog # 470332-592
Supplier:  
Fischertechnik TXT 4.0 Base Set
Fischertechnik TXT 4.0 Base Set
Fischertechnik TXT 4.0 base set
Discontinued
Product Details & Documents
Measurement, control, regulation, and programming for middle/high school and beyond.
  • Acts as the core set for all fischertechnik robotic add-on products
  • Includes downloadable teaching and activity material (free download)
Main topics include robotics and programming (graphical and text-based), actuators, analog and digital sensors, measurement, control, data transmission, coding-decoding, and image processing. Set included a robotics TXT 4.0 controller, robo pro coding software, accu set, 2 encoder motors, USB camera, ultrasonic sensor, track sensor, 2 push button switches, 2 LEDs, and a photo-transistor.
Spécifications
  • Désignation:
    Fischertechnik TXT 4.0 base set