Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Cutters, Carbide Insert, Excelta | Avantor

Cutters, Carbide Insert, Excelta

Cutters, small round head, hard wire, carbide inserts, cleanroom safe, Stainless steel, 112.5 mm (4.5"), 12°
Catalog # 76471-616
Supplier:   Excelta
Fournisseur #:  2899EHT-CR
Cutters, Carbide Insert, Excelta
Catalog # 76471-616
Supplier:  Excelta
Cutters, Carbide Insert, Excelta
Cutters, Carbide Insert, Excelta

Cutters, small round head, hard wire, carbide inserts, cleanroom safe, Stainless steel, 112.5 mm (4.5"), 12°

Price
$154.72
Log in pour la disponibilité
Each
Product Details & Documents

This compound action cutter is ideal for cutting small hard wire.

  • Carbide insert cutting edges for tough hard wire
  • Compound-Action means less pressure on hands
  • Dual leaf springs for ease of use
  • Stainless steel shank for corrosion resistants

These multi-purpose medium round hard wire cutters feature stainless steel construction for corrosion resistance and carbide inserts for maximum blade life. Hard wire cutting capacity is 0.010 to 0.020". The grips are soft cushioned foam and have a surface resistivity of 10¹⁰ ohms/sq. Interestingly, the 149AT is a compound-action cutter, which requires much less force to operate.

Spécifications
  • Désignation:
    Cutters, small round head, hard wire, carbide inserts, cleanroom safe
  • Matière:
    Stainless steel
  • Curvature:
    12°
  • Overall Length:
    112.5 mm (4.5")
Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.