Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Small Pliers, Excelta | Avantor

Small Pliers, Excelta

0.340 W×0.235 D", 4½", 0.020 W×0.035 D", Needle nose
Catalog # 76471-518
Supplier:   Excelta
Fournisseur #:  47I
Small Pliers, Excelta
Catalog # 76471-518
Supplier:  Excelta
Small Pliers, Excelta
Small Pliers, Excelta

0.340 W×0.235 D", 4½", 0.020 W×0.035 D", Needle nose

Price
$153.24
Log in pour la disponibilité
Each
Product Details & Documents

Three star quality small pliers made of carbon steel with flame hardened tips and feature dual leaf springs for smooth action.The grips are soft cushioned foam and are ESD safe.

Spécifications
  • Longueur:
    4½"
  • Taille:
    0.340 W×0.235 D"
  • Tip Size:
    0.020 W×0.035 D"
  • Tip Style:
    Needle nose