Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Small Pliers, Excelta

Small Pliers, Excelta

0.355 W×0.235 D", 4½", 0.020 W×0.039 D", Chain nose
Catalog # 76471-516
Supplier:   Excelta
Supplier #:  44I
Small Pliers, Excelta
Catalog # 76471-516
Supplier:  Excelta
Small Pliers, Excelta
Small Pliers, Excelta

0.355 W×0.235 D", 4½", 0.020 W×0.039 D", Chain nose

Price
$153.24
Log in for availability
Each
Product Details & Documents

Three star quality small pliers made of carbon steel with flame hardened tips and feature dual leaf springs for smooth action.The grips are soft cushioned foam and are ESD safe.

Specifications
  • Longueur:
    4½"
  • Taille:
    0.355 W×0.235 D"
  • Tip Size:
    0.020 W×0.039 D"
  • Tip Style:
    Chain nose
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.