Ward's® Basalt (Flood) | Avantor
Ward's® Basalt (Flood)
Basalt (Flood) thin section
Catalog # 470331-426
Supplier:   Avantor
Ward's® Basalt (Flood)
Catalog # 470331-426
Supplier:  Avantor
Ward's® Basalt (Flood)
Ward's® Basalt (Flood)
Basalt (Flood) thin section
Price
$96.59
Log in pour la disponibilité
Each
Product Details & Documents
Classic, dark; from the Columbia Plateau.
  • Collected in Washington
  • Multiple sizes and pack options to fit specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All Specimen dimensions are considered an average size range.
Spécifications
  • Désignation:
    Basalt (Flood) thin section
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.