Miltex® Forceps, Integra | Avantor
Miltex® Forceps, Integra
Tissue forceps, 1×2 teeth, serrated handles, 129 mm (5⅛")
Catalog # 76470-896
Supplier:   Miltex
Fournisseur #:  6-42
Miltex® Forceps, Integra
Catalog # 76470-896
Supplier:  Miltex
Miltex® Forceps, Integra
Miltex® Forceps, Integra
Tissue forceps, 1×2 teeth, serrated handles, 129 mm (5⅛")
Price
$71.44
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Integra Miltex® forceps can be used for delicate procedural requirements as well as routine yet important surgical needs.
  • Sterile
  • Includes common instrument patterns for routine surgical procedures
It may be instrumentation for exacting plastic surgery with just the right handle or a simple 'pick-up'. Miltex® is able to fulfill either demand with just the right instrument.
Spécifications
  • Désignation:
    Tissue forceps, 1×2 teeth, serrated handles
  • Longueur:
    129 mm (5⅛")