Miltex® Scissors, Integra
Product Details & Documents
Integra Miltex recognizes and meets a variety of clinical needs in the careful construction of these surgical scissors.
Whether dissecting, cutting tissue, suture or dressing material or performing any number of important functions, our finely crafted scissors perform as intended for the surgical task.
Specifications
- Désignation:Operating scissors, SuperCut, one serrated blade
- Longueur:145 mm (5¾")
- Curvature:Curved
- Blade Type:Sharp-blunt points