Miltex® Scissors, Integra | Avantor
Miltex® Scissors, Integra
Iris scissors, SuperCut, 116 mm (4⅝"), Sharp/sharp, Straight
Catalog # 76470-888
Supplier:   Miltex
Fournisseur #:  5-SC-304
Miltex® Scissors, Integra
Catalog # 76470-888
Supplier:  Miltex
Miltex® Scissors, Integra
Iris scissors, SuperCut, 116 mm (4⅝"), Sharp/sharp, Straight
Price
373,77 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Integra Miltex recognizes and meets a variety of clinical needs in the careful construction of these surgical scissors.
Whether dissecting, cutting tissue, suture or dressing material or performing any number of important functions, our finely crafted scissors perform as intended for the surgical task.
Spécifications
  • Désignation:
    Iris scissors, SuperCut
  • Longueur:
    116 mm (4⅝")
  • Curvature:
    Straight
  • Blade Type:
    Sharp/sharp