Tungsten Carbide Cutters, Electron Microscopy Sciences
Product Details & Documents
These cutters and pliers are designed for use in applications where strength, wear resistance, and hardness are a primary concern.
- Magnetizable
- Harden by heat treatment
- ESD safe
Tungsten carbide and high precision cutters are made from carbon steel, with a maximum hardness of 68 HRC. They offer a moderate resistance to corrosion, and are ideal for cutting and bending single or multiple filars, and for lateral or internal cuts.
ESD epoxy-coated cutting tweezers have a poor resisance to corrosion. They are often used as tweezers in medical device manufacturing, and as a cutting tool in the electricity industry.
Documents
Certificates
Recherche par numéro de lotSpécifications
- Désignation:High precision cutter, style ES5152
- Jaw Type:Large oval
- Overall Length:120 mm