Tungsten Carbide Cutters, Electron Microscopy Sciences | Avantor
Tungsten Carbide Cutters, Electron Microscopy Sciences
High precision cutter, style ES5351, 120 mm, Large tapered
Catalog # 76458-606
Fournisseur #:  9506
Tungsten Carbide Cutters, Electron Microscopy Sciences
Catalog # 76458-606
Tungsten Carbide Cutters, Electron Microscopy Sciences
Tungsten Carbide Cutters, Electron Microscopy Sciences
High precision cutter, style ES5351, 120 mm, Large tapered
Price
$277.26
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These cutters and pliers are designed for use in applications where strength, wear resistance, and hardness are a primary concern.
  • Magnetizable
  • Harden by heat treatment
  • ESD safe
Tungsten carbide and high precision cutters are made from carbon steel, with a maximum hardness of 68 HRC. They offer a moderate resistance to corrosion, and are ideal for cutting and bending single or multiple filars, and for lateral or internal cuts.
ESD epoxy-coated cutting tweezers have a poor resisance to corrosion. They are often used as tweezers in medical device manufacturing, and as a cutting tool in the electricity industry.
Documents
Spécifications
  • Désignation:
    High precision cutter, style ES5351
  • Jaw Type:
    Large tapered
  • Overall Length:
    120 mm