Nitrile Examination Gloves, Textured, Electron Microscopy Sciences | Avantor
Nitrile Examination Gloves, Textured, Electron Microscopy Sciences
L, Brown
Catalog # 76458-560
Nitrile Examination Gloves, Textured, Electron Microscopy Sciences
Catalog # 76458-560
Supplier:  
Nitrile Examination Gloves, Textured, Electron Microscopy Sciences
Nitrile Examination Gloves, Textured, Electron Microscopy Sciences
L, Brown
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
Product Details & Documents
Gloves are made from 100% synthetic nitrile that contains no latex. They offer superior textile strength and are puncture-resistant when compared to other latex or vinyl gloves of similar thickness.
  • Textured finish provides a positive grip on wet and/or dry surfaces
  • Excellent textile strength
  • Puncture-resistant
  • Latex-free
  • Powder-free
These powder-free gloves will provide the user with superior comfort, excellent flexibility, and dexterity. The nitrile compound provides a broad range of hand protection against accidental chemical splashes. There are no natural rubber protein allergens as is present in natural latex rubber gloves.
Spécifications
  • Glove Color:
    Brown
  • Taille du gant:
    L
  • Powdered/Powder-free:
    Powder-free
  • Cat. No.:
    76458-560
  • Disposable / reusable:
    Disposable
  • Latex/Latex-free:
    Latex-free
  • Sterility:
    Non sterile
  • Finish:
    Textured