Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Ward's OneRack

Ward's® OneRack (blue/clear), 5 - 50 ml, 12.2×4.2×3"
Catalog # 470330-002
Supplier:   Heathrow Scientific
 :  120817W
Ward's OneRack
Catalog # 470330-002
Ward's OneRack
Ward's OneRack

Ward's® OneRack (blue/clear), 5 - 50 ml, 12.2×4.2×3"

Price
$80.60
Log in pour la disponibilité
Each Case of 50
Product Details & Documents

The most flexible tube rack designed for use with a multitude of tube diameters and heights.

  • Interchangeable inserts accommodate multiple sized tubes
  • Inserts can be pivoted as needed
  • Open design allows visibility of tube graduations when filling
  • Teardrop shaped wells allow better retention of tubes

The Ward's® OneRack and its interchangeable shelves can accommodate virtually any brand tube (10 to 30 mm) from 5 to 50 ml volume, even if they have varying heights. Inserts can be pivoted in either an upper or lower position to securely accommodate the size and height of tube being used.

Spécifications
  • Désignation:
    Ward's® OneRack (blue/clear)
  • L×W×H:
    12.2×4.2×3"
  • For Tubes:
    5 - 50 ml