Graphene Oxide TEM Support Films, Electron Microscopy Sciences
Graphene Oxide TEM Support Films, Electron Microscopy Sciences
300, Copper
Catalog # 76439-712
Supplier #:  GOLC300CU50
Graphene Oxide TEM Support Films, Electron Microscopy Sciences
Catalog # 76439-712
Graphene Oxide TEM Support Films, Electron Microscopy Sciences
Graphene Oxide TEM Support Films, Electron Microscopy Sciences
300, Copper
Price
$2,007.44
Log in pour la disponibilité
Pack of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Graphene Oxide (GO) support film is a super thin (<1 nm), naturally hydrophilic layer placed over the Holey, Lacey or Quantifoil support film on copper or gold grids.
  • Works well with Holey Carbon, Lacey Carbon and Quantifoil grid types, effectively spanning the gaps
  • Less expensive to produce due to complexity of graphene manufacturing
  • Better background contrast than graphene, results in higher resolution
Pre-treatment of GO Support Films is unnecessary. By default, the hydrophilic surface spreads particles evenly across the grid. A hydrophobic surface can be achieved by heating in the air. Note: plasma cleaning or glow discharge will damage the support film.
Spécifications
  • Grid Type:
    Copper
  • Mesh:
    300