Graphene Oxide TEM Support Films, Electron Microscopy Sciences
300, Copper
  76439-700
 :  GOHC300CU10
Graphene Oxide TEM Support Films, Electron Microscopy Sciences
  76439-700
Graphene Oxide TEM Support Films, Electron Microscopy Sciences
Graphene Oxide TEM Support Films, Electron Microscopy Sciences
300, Copper
$489.05
Pack of 10
Graphene Oxide (GO) support film is a super thin (<1 nm), naturally hydrophilic layer placed over the Holey, Lacey or Quantifoil support film on copper or gold grids.
  • Works well with Holey Carbon, Lacey Carbon and Quantifoil grid types, effectively spanning the gaps
  • Less expensive to produce due to complexity of graphene manufacturing
  • Better background contrast than graphene, results in higher resolution
Pre-treatment of GO Support Films is unnecessary. By default, the hydrophilic surface spreads particles evenly across the grid. A hydrophobic surface can be achieved by heating in the air. Note: plasma cleaning or glow discharge will damage the support film.
  • Grid Type:
    Copper
  • Mesh:
    300